Learn More
Eurofins Discoverx CHO-K1 F2RL1 Gq Cell Line, 2 vials. (95-1006C2)
Catalog No. 502004037
Encompass
The F2RL1 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serine proteases, SLIGRL-NH2) based activation of F2RL1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses F2RL1, a naturally Gq coupled GPCR. Upon activation of F2RL1, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4037 Supplier Eurofins Discoverx Supplier No. 951006C2
Add to Cart
Add to Cart
Request a Quote
The F2RL1 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serine proteases, SLIGRL-NH2) based activation of F2RL1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses F2RL1, a naturally Gq coupled GPCR. Upon activation of F2RL1, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.