Learn More
Eurofins Discoverx PathHunter CHO-K1 FFAR2 β-Arrestin Cell Line, 2 vials. (93-1005C2)
Catalog No. 502001818
Encompass
The PathHunter FFAR2 (GPR43) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acid) based activation of FFAR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged FFAR2 and EA-tagged β-Arrestin-2. Activation of FFAR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1818 Supplier Eurofins Discoverx Supplier No. 931005C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter FFAR2 (GPR43) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acid) based activation of FFAR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged FFAR2 and EA-tagged β-Arrestin-2. Activation of FFAR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.