Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 FPR1 Gi Cell Line, 2 vials. (95-0139C2)
Supplier:  Eurofins Discoverx 950139C2
Encompass
The cAMP Hunter FPR1 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of FPR1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type FPR1, a naturally Gi/Go coupled GPCR. Upon activation of FPR1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3650
Add to Cart
Request a Quote