Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 GABBR1-GABBR2 Gi Cell Line, 2 vials. (95-0165C2)
Catalog No. 502003930
Encompass
The cAMP Hunter GABBR1-GABBR2 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GABA) based activation of GABBR1/GABBR2 GPCR heterodimer activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GABBR1, a naturally Gi/Go coupled GPCR, alongside its partner subunit GABBR2. Upon activation of GABBR1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3930 Supplier Eurofins Discoverx Supplier No. 950165C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter GABBR1-GABBR2 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GABA) based activation of GABBR1/GABBR2 GPCR heterodimer activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GABBR1, a naturally Gi/Go coupled GPCR, alongside its partner subunit GABBR2. Upon activation of GABBR1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.