Learn More
Eurofins Discoverx PathHunter CHO-K1 GHSR1b β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0354C2A)
Catalog No. 502000332
Encompass
The PathHunter GHSR1b cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ghrelin) based activation of GHSR1b GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR1b and EA-tagged β-Arrestin-2. Activation of GHSR1b stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0332 Supplier Eurofins Discoverx Supplier No. 930354C2A
Add to Cart
Add to Cart
Request a Quote
The PathHunter GHSR1b cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ghrelin) based activation of GHSR1b GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR1b and EA-tagged β-Arrestin-2. Activation of GHSR1b stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.