Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 GIPR Gs Cell Line, 2 vials. (95-0146C2)
Catalog No. 502003677
Encompass
The cAMP Hunter GIPR Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GIP) based activation of GIPR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GIPR, a naturally Gs coupled GPCR. Upon activation of GIPR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3677 Supplier Eurofins Discoverx Supplier No. 950146C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter GIPR Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GIP) based activation of GIPR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GIPR, a naturally Gs coupled GPCR. Upon activation of GIPR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.