Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 GPR109A Gi Cell Line, 2 vials. (95-0122C2)
Catalog No. 502003317
Encompass
The cAMP Hunter GPR109A Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. nicotinic acid) based activation of GPR109A GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GPR109A, a naturally Gi/Go coupled GPCR. Upon activation of GPR109A, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3317 Supplier Eurofins Discoverx Supplier No. 950122C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter GPR109A Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. nicotinic acid) based activation of GPR109A GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GPR109A, a naturally Gi/Go coupled GPCR. Upon activation of GPR109A, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.