Learn More
Eurofins Discoverx CHO-K1 GPR17 Gq Cell Line, 2 vials. (95-1022C2)
Catalog No. 502004123
Encompass
The GPR17 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MDL 29,951) based activation of GPR17 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GPR17, a naturally Gq coupled GPCR. Upon activation of GPR17, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4123 Supplier Eurofins Discoverx Supplier No. 951022C2
Add to Cart
Add to Cart
Request a Quote
The GPR17 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MDL 29,951) based activation of GPR17 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GPR17, a naturally Gq coupled GPCR. Upon activation of GPR17, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.