Learn More
Eurofins Discoverx PathHunter CHO-K1 GPR182 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0754C2)
Supplier:  Eurofins Discoverx 930754C2
Encompass
The PathHunter GPR182 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR182 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR182 and EA-tagged β-Arrestin-2. Activation of GPR182 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1249
Only null left
Add to Cart
Request a Quote