Learn More
Eurofins Discoverx PathHunter CHO-K1 GPR32 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0321C2A)
Catalog No. 502000273
Encompass
The PathHunter GPR32 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR32 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR32 and EA-tagged β-Arrestin-2. Activation of GPR32 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0273 Supplier Eurofins Discoverx Supplier No. 930321C2A
Add to Cart
Add to Cart
Request a Quote
The PathHunter GPR32 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR32 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR32 and EA-tagged β-Arrestin-2. Activation of GPR32 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.