Learn More
Eurofins Discoverx PathHunter CHO-K1 GPR50 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0648C2)
Supplier:  Eurofins Discoverx 930648C2
Encompass
The PathHunter GPR50 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR50 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR50 and EA-tagged β-Arrestin-2. Activation of GPR50 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1024
Add to Cart
Request a Quote