Learn More
Eurofins Discoverx PathHunter CHO-K1 GPR84 β-Arrestin Cell Line, 2 vials. (93-0647C2)
Catalog No. 502001023
Encompass
The PathHunter GPR84 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of GPR84 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR84 and EA-tagged β-Arrestin-2. Activation of GPR84 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1023 Supplier Eurofins Discoverx Supplier No. 930647C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter GPR84 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of GPR84 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR84 and EA-tagged β-Arrestin-2. Activation of GPR84 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.