Learn More
Eurofins Discoverx PathHunter CHO-K1 HCRTR2 β-Arrestin Cell Line, 2 vials. (93-0427C2)
Supplier:  Eurofins Discoverx 930427C2
Encompass
The PathHunter HCRTR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. orexin A) based activation of HCRTR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HCRTR2 and EA-tagged β-Arrestin-2. Activation of HCRTR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0521
Add to Cart
Request a Quote