Learn More
Eurofins Discoverx PathHunter CHO-K1 HRH2 β-Arrestin Cell Line, 2 vials. (93-0208C2)
Catalog No. 501999953
Encompass
The PathHunter HRH2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. histamine) based activation of HRH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HRH2 and EA-tagged β-Arrestin-2. Activation of HRH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9953 Supplier Eurofins Discoverx Supplier No. 930208C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter HRH2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. histamine) based activation of HRH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HRH2 and EA-tagged β-Arrestin-2. Activation of HRH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.