Learn More
Eurofins Discoverx PathHunter CHO-K1 HTR1A β-Arrestin Cell Line, 2 vials. (93-0696C2)
Catalog No. 502001135
Encompass
The PathHunter HTR1A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR1A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1A and EA-tagged β-Arrestin-2. Activation of HTR1A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1135 Supplier Eurofins Discoverx Supplier No. 930696C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter HTR1A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR1A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1A and EA-tagged β-Arrestin-2. Activation of HTR1A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.