Learn More
Eurofins Discoverx PathHunter CHO-K1 LGR5 (GPR49) β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0526C2A)
Supplier:  Eurofins Discoverx 930526C2A
Encompass
The PathHunter LGR5 (GPR49) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. R-spondins) based activation of LGR5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged LGR5 and EA-tagged β-Arrestin-2. Activation of LGR5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0716
Only null left
Add to Cart
Request a Quote