Learn More
Eurofins Discoverx PathHunter CHO-K1 LHCGR β-Arrestin Cell Line, 2 vials. (93-0957C2)
Catalog No. 502001663
Encompass
The PathHunter LHCGR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. luteinizing hormone) based activation of LHCGR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged LHCGR and EA-tagged β-Arrestin-2. Activation of LHCGR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1663 Supplier Eurofins Discoverx Supplier No. 930957C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter LHCGR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. luteinizing hormone) based activation of LHCGR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged LHCGR and EA-tagged β-Arrestin-2. Activation of LHCGR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.