Learn More
Eurofins Discoverx PathHunter CHO-K1 LTB4R β-Arrestin Cell Line, 2 vials. (93-0292C2)
Catalog No. 502000186
Encompass
The PathHunter LTB4R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. leukotriene B4) based activation of LTB4R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged LTB4R and EA-tagged β-Arrestin-2. Activation of LTB4R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0186 Supplier Eurofins Discoverx Supplier No. 930292C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter LTB4R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. leukotriene B4) based activation of LTB4R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged LTB4R and EA-tagged β-Arrestin-2. Activation of LTB4R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.