Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 LTB4R Gi Cell Line, 2 vials. (95-0085C2)
Catalog No. 502002838
Encompass
The cAMP Hunter LTB4R Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. leukotriene B4) based activation of LTB4R GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type LTB4R, a naturally Gi/Go coupled GPCR. Upon activation of LTB4R, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-2838 Supplier Eurofins Discoverx Supplier No. 950085C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter LTB4R Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. leukotriene B4) based activation of LTB4R GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type LTB4R, a naturally Gi/Go coupled GPCR. Upon activation of LTB4R, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.