Learn More
Eurofins Discoverx PathHunter CHO-K1 mADRA1B β-Arrestin Cell Pool, 2 vials. (93-0595P2)
Supplier:  Eurofins Discoverx 930595P2
Encompass
The PathHunter mADRA1B cell pool is a stable pool, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of mADRA1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mADRA1B and EA-tagged β-Arrestin-2. Activation of mADRA1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0917
Add to Cart
Request a Quote