Learn More
Eurofins Discoverx PathHunter CHO-K1 mCCKAR β-Arrestin Cell Pool, 2 vials. (93-0598P2)
Catalog No. 502000920
Encompass
The PathHunter mCCKAR cell pool is a stable, engineered cell pool for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cholecystokinin) based activation of mCCKAR GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mCCKAR and EA-tagged β-Arrestin-2. Activation of mCCKAR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0920 Supplier Eurofins Discoverx Supplier No. 930598P2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mCCKAR cell pool is a stable, engineered cell pool for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cholecystokinin) based activation of mCCKAR GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mCCKAR and EA-tagged β-Arrestin-2. Activation of mCCKAR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.