Learn More
Eurofins Discoverx PathHunter CHO-K1 mCCR8 β-Arrestin Cell Line, 2 vials. (93-0556C2)
Catalog No. 502000785
Encompass
The PathHunter mCCR8 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL1) based activation of mCCR8 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCCR8 and EA-tagged β-Arrestin-2. Activation of mCCR8 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0785 Supplier Eurofins Discoverx Supplier No. 930556C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mCCR8 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL1) based activation of mCCR8 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCCR8 and EA-tagged β-Arrestin-2. Activation of mCCR8 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.