Learn More
Eurofins Discoverx PathHunter CHO-K1 mCXCR3 β-Arrestin Cell Line, 2 vials. (93-0553C2)
Catalog No. 502000781
Encompass
The PathHunter mCXCR3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL11/I-TAC) based activation of mCXCR3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCXCR3 and EA-tagged β-Arrestin-2. Activation of mCXCR3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0781 Supplier Eurofins Discoverx Supplier No. 930553C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mCXCR3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL11/I-TAC) based activation of mCXCR3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCXCR3 and EA-tagged β-Arrestin-2. Activation of mCXCR3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.