Learn More
Eurofins Discoverx PathHunter CHO-K1 mGLP1R β-Arrestin-1 Cell Line, 2 vials. (93-0974C2)
Catalog No. 502001721
Encompass
The PathHunter mGLP1R (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. exendin-4) based activation of mGLP1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGLP1R and EA-tagged β-Arrestin-1. Activation of mGLP1R stimulates the recruitment of β-Arrestin-1 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1721 Supplier Eurofins Discoverx Supplier No. 930974C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mGLP1R (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. exendin-4) based activation of mGLP1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGLP1R and EA-tagged β-Arrestin-1. Activation of mGLP1R stimulates the recruitment of β-Arrestin-1 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.