Learn More
Eurofins Discoverx PathHunter CHO-K1 mGPR84 β-Arrestin Cell Line, 2 vials. (93-1083C2)
Catalog No. 502002009
Encompass
The PathHunter mGPR84 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of mGPR84 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGPR84 and EA-tagged β-Arrestin-2. Activation of mGPR84 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-2009 Supplier Eurofins Discoverx Supplier No. 931083C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mGPR84 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of mGPR84 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGPR84 and EA-tagged β-Arrestin-2. Activation of mGPR84 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.