Learn More
Eurofins Discoverx PathHunter CHO-K1 mHTR2A β-Arrestin Cell Line, 2 vials. (93-0567C2)
Catalog No. 502000859
Encompass
The PathHunter mHTR2A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin/5-HT) based activation of mHTR2A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mHTR2A and EA-tagged β-Arrestin-2. Activation of mHTR2A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0859 Supplier Eurofins Discoverx Supplier No. 930567C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mHTR2A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin/5-HT) based activation of mHTR2A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mHTR2A and EA-tagged β-Arrestin-2. Activation of mHTR2A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.