Learn More
Eurofins Discoverx PathHunter CHO-K1 mMC3R β-Arrestin Cell Pool, 2 vials. (93-0642P2)
Catalog No. 502001017
Encompass
The PathHunter mMC3R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-MSH) based activation of mMC3R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mMC3R and EA-tagged β-Arrestin-2. Activation of mMC3R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1017 Supplier Eurofins Discoverx Supplier No. 930642P2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mMC3R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-MSH) based activation of mMC3R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mMC3R and EA-tagged β-Arrestin-2. Activation of mMC3R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.