Learn More
Eurofins Discoverx PathHunter CHO-K1 mMC4R β-Arrestin Cell Pool, 2 vials. (93-0608P2)
Catalog No. 502000936
Encompass
The PathHunter mMC4R cell pool is a stable, engineered cell pool for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-MSH) based activation of mMC4R GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mMC4R and EA-tagged β-Arrestin-2. Activation of mMC4R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0936 Supplier Eurofins Discoverx Supplier No. 930608P2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mMC4R cell pool is a stable, engineered cell pool for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-MSH) based activation of mMC4R GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mMC4R and EA-tagged β-Arrestin-2. Activation of mMC4R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.