Learn More
Eurofins Discoverx PathHunter CHO-K1 mNPY2R β-Arrestin Cell Line, 2 vials. (93-0569C2)
Catalog No. 502000860
Encompass
The PathHunter mNPY2R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of mNPY2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mNPY2R and EA-tagged β-Arrestin-2. Activation of mNPY2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0860 Supplier Eurofins Discoverx Supplier No. 930569C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mNPY2R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of mNPY2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mNPY2R and EA-tagged β-Arrestin-2. Activation of mNPY2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.