Learn More
Eurofins Discoverx PathHunter CHO-K1 mPTGIR β-Arrestin Cell Pool, 2 vials. (93-0615P2)
Catalog No. 502000952
Encompass
The PathHunter mPTGIR cell pool is a stable, engineered cell pool for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell pool enables assessment of ligand (e.g. prostaglandin I2, beraprost) based activation of mPTGIR GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mPTGIR and EA-tagged β-Arrestin-2. Activation of mPTGIR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0952 Supplier Eurofins Discoverx Supplier No. 930615P2
Add to Cart
Add to Cart
Request a Quote
The PathHunter mPTGIR cell pool is a stable, engineered cell pool for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell pool enables assessment of ligand (e.g. prostaglandin I2, beraprost) based activation of mPTGIR GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mPTGIR and EA-tagged β-Arrestin-2. Activation of mPTGIR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.