Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 mRXFP1 Gs Cell Line, 2 vials. (95-0180C2)
Supplier:  Eurofins Discoverx 950180C2
Encompass
The cAMP Hunter mRXFP1 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. relaxin-2) based activation of mRXFP1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type mRXFP1, a naturally Gs coupled GPCR. Upon activation of mRXFP1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3992
Add to Cart
Request a Quote