Learn More
Eurofins Discoverx PathHunter CHO-K1 mTACR1 β-Arrestin Cell Pool, 2 vials. (93-0618P2)
Supplier:  Eurofins Discoverx 930618P2
Encompass
The PathHunter mTACR1 cell pool is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell pool enables assessment of ligand (e.g. substance P) based activation of mTACR1 GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mTACR1 and EA-tagged β-Arrestin-2. Activation of mTACR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0957
Add to Cart
Request a Quote