Learn More
Eurofins Discoverx PathHunter CHO-K1 MTNR1B β-Arrestin Cell Line, 2 vials. (93-0630C2)
Catalog No. 502000980
Encompass
The PathHunter MTNR1B cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. melatonin) based activation of MTNR1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MTNR1B and EA-tagged β-Arrestin-2. Activation of MTNR1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0980 Supplier Eurofins Discoverx Supplier No. 930630C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter MTNR1B cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. melatonin) based activation of MTNR1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MTNR1B and EA-tagged β-Arrestin-2. Activation of MTNR1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.