Learn More
Eurofins Discoverx PathHunter CHO-K1 NPR1 Functional Assay, 2 vials. (93-0804C2)
Catalog No. 502001325
Encompass
The PathHunter CHO-K1 NPR1 Functional Assay is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting NPR1. This cell line enables assessment of ligand (e.g. ANP) based stimulation of receptor activity via detection of resulting increases in the levels of intracellular cGMP. This product uses EFC technology. The included cell line overexpresses untagged NPR1 Upon activation of the pathway, the increase in levels of intracellular cGMP leads to production of increased EFC signal when the cGMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1325 Supplier Eurofins Discoverx Supplier No. 930804C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter CHO-K1 NPR1 Functional Assay is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting NPR1. This cell line enables assessment of ligand (e.g. ANP) based stimulation of receptor activity via detection of resulting increases in the levels of intracellular cGMP. This product uses EFC technology. The included cell line overexpresses untagged NPR1 Upon activation of the pathway, the increase in levels of intracellular cGMP leads to production of increased EFC signal when the cGMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.