Learn More
Eurofins Discoverx PathHunter CHO-K1 NPY1R β-Arrestin Cell Line, 2 vials. (93-0397C2)
Catalog No. 502000436
Encompass
The PathHunter NPY1R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of NPY1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged NPY1R and EA-tagged β-Arrestin-2. Activation of NPY1R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0436 Supplier Eurofins Discoverx Supplier No. 930397C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter NPY1R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of NPY1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged NPY1R and EA-tagged β-Arrestin-2. Activation of NPY1R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.