Learn More
Eurofins Discoverx PathHunter CHO-K1 OXER1 β-Arrestin Cell Line, 2 vials. (93-0867C2)
Catalog No. 502001451
Encompass
The PathHunter OXER1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. 5-oxo-ETE) based activation of OXER1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged OXER1 and EA-tagged β-Arrestin-2. Activation of OXER1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1451 Supplier Eurofins Discoverx Supplier No. 930867C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter OXER1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. 5-oxo-ETE) based activation of OXER1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged OXER1 and EA-tagged β-Arrestin-2. Activation of OXER1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.