Learn More
Eurofins Discoverx PathHunter CHO-K1 OXGR1 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0693C2)
Catalog No. 502001126
Encompass
The PathHunter OXGR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. alpha-ketoglutaric acid) based activation of OXGR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged OXGR1 and EA-tagged β-Arrestin-2. Activation of OXGR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1126 Supplier Eurofins Discoverx Supplier No. 930693C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter OXGR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. alpha-ketoglutaric acid) based activation of OXGR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged OXGR1 and EA-tagged β-Arrestin-2. Activation of OXGR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.