Learn More
Eurofins Discoverx PathHunter CHO-K1 P2RY8 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0635C2)
Catalog No. 502000993
Encompass
The PathHunter P2RY8 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. UTP) based activation of P2RY8 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged P2RY8 and EA-tagged β-Arrestin-2. Activation of P2RY8 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0993 Supplier Eurofins Discoverx Supplier No. 930635C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter P2RY8 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. UTP) based activation of P2RY8 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged P2RY8 and EA-tagged β-Arrestin-2. Activation of P2RY8 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.