Learn More
Eurofins Discoverx PathHunter CHO-K1 PROKR1 β-Arrestin Cell Line, 2 vials. (93-0303C2)
Catalog No. 502000210
Encompass
The PathHunter PROKR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prokinectin, EG-VEGF) based activation of PROKR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PROKR1 and EA-tagged β-Arrestin-2. Activation of PROKR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0210 Supplier Eurofins Discoverx Supplier No. 930303C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter PROKR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prokinectin, EG-VEGF) based activation of PROKR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PROKR1 and EA-tagged β-Arrestin-2. Activation of PROKR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.