Learn More
Eurofins Discoverx PathHunter CHO-K1 PTAFR β-Arrestin Cell Line, 2 vials. (93-0236C2)
Catalog No. 502000034
Encompass
The PathHunter PTAFR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. PAF) based activation of PTAFR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTAFR and EA-tagged β-Arrestin-2. Activation of PTAFR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0034 Supplier Eurofins Discoverx Supplier No. 930236C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter PTAFR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. PAF) based activation of PTAFR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTAFR and EA-tagged β-Arrestin-2. Activation of PTAFR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.