Learn More
Eurofins Discoverx CHO-K1 PTAFR Gq Cell Line, 2 vials. (95-0091C2)
Catalog No. 502002934
Encompass
The PTAFR Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. PAF) based activation of PTAFR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTAFR, a naturally Gq coupled GPCR. Upon activation of PTAFR, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-2934 Supplier Eurofins Discoverx Supplier No. 950091C2
Add to Cart
Add to Cart
Request a Quote
The PTAFR Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. PAF) based activation of PTAFR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTAFR, a naturally Gq coupled GPCR. Upon activation of PTAFR, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.