Learn More
Eurofins Discoverx CHO-K1 PTGFR Gq Cell Line, 2 vials. (95-1009C2)
Supplier:  Eurofins Discoverx 951009C2
Encompass
The PTGFR Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin F2a) based activation of PTGFR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTGFR, a naturally Gq coupled GPCR. Upon activation of PTGFR, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4058
Add to Cart
Request a Quote