Learn More
Eurofins Discoverx PathHunter CHO-K1 PTGIR β-Arrestin Cell Line, 2 vials. (93-0305C2)
Catalog No. 502000220
Encompass
The PathHunter PTGIR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin I2, beraprost) based activation of PTGIR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGIR and EA-tagged β-Arrestin-2. Activation of PTGIR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0220 Supplier Eurofins Discoverx Supplier No. 930305C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter PTGIR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin I2, beraprost) based activation of PTGIR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGIR and EA-tagged β-Arrestin-2. Activation of PTGIR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.