Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 PTGIR Gs Cell Line, 2 vials. (95-0051C2)
Catalog No. 502002471
Encompass
The cAMP Hunter PTGIR Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin I2, beraprost) based activation of PTGIR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTGIR, a naturally Gs coupled GPCR. Upon activation of PTGIR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-2471 Supplier Eurofins Discoverx Supplier No. 950051C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter PTGIR Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin I2, beraprost) based activation of PTGIR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTGIR, a naturally Gs coupled GPCR. Upon activation of PTGIR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.