Learn More
Eurofins Discoverx PathHunter CHO-K1 rDRD2L (Long Isoform) β-Arrestin Cell Line, 2 vials. (93-0785C2)
Catalog No. 502001311
Encompass
The PathHunter rDRD2L cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of rDRD2L GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rDRD2L and EA-tagged β-Arrestin-2. Activation of rDRD2L stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1311 Supplier Eurofins Discoverx Supplier No. 930785C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter rDRD2L cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of rDRD2L GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rDRD2L and EA-tagged β-Arrestin-2. Activation of rDRD2L stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.