Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 rDRD2L (Long Isoform) Gi Cell Line, 2 vials. (95-0130C2)
Catalog No. 502003587
Encompass
The cAMP Hunter rDRD2L Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine, pergolide) based activation of rDRD2L GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type rDRD2L, a naturally Gi/Go coupled GPCR. Upon activation of rDRD2L, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3587 Supplier Eurofins Discoverx Supplier No. 950130C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter rDRD2L Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine, pergolide) based activation of rDRD2L GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type rDRD2L, a naturally Gi/Go coupled GPCR. Upon activation of rDRD2L, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.