Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 rDRD2S (Short Isoform) Gi Cell Line, 2 vials. (95-0129C2)
Supplier:  Eurofins Discoverx 950129C2
Encompass
The cAMP Hunter rDRD2S Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine, pergolide) based activation of rDRD2S GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type rDRD2S, a naturally Gi/Go coupled GPCR. Upon activation of rDRD2S, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3582
Only null left
Add to Cart
Request a Quote