Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 rTAAR1 Gs Cell Line, 2 vials. (95-0178C2)
Catalog No. 502003972
Encompass
The cAMP Hunter rTAAR1 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. p-tyramine) based activation of rTAAR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type rTAAR1, a naturally Gs coupled GPCR. Upon activation of rTAAR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3972 Supplier Eurofins Discoverx Supplier No. 950178C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter rTAAR1 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. p-tyramine) based activation of rTAAR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type rTAAR1, a naturally Gs coupled GPCR. Upon activation of rTAAR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.