Learn More
Eurofins Discoverx PathHunter CHO-K1 RXFP3 β-Arrestin Cell Line, 2 vials. (93-0692C2)
Catalog No. 502001123
Encompass
The PathHunter RXFP3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. relaxin-3) based activation of RXFP3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged RXFP3 and EA-tagged β-Arrestin-2. Activation of RXFP3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1123 Supplier Eurofins Discoverx Supplier No. 930692C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter RXFP3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. relaxin-3) based activation of RXFP3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged RXFP3 and EA-tagged β-Arrestin-2. Activation of RXFP3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.