Learn More
Eurofins Discoverx PathHunter CHO-K1 RXRγ Protein Interaction Cell Line, 2 vials. (93-0327C2)
Supplier:  Eurofins Discoverx 930327C2
Encompass
The PathHunter RXRγ Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. retinoic acid) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. The included cell line overexpresses ProLink-tagged RXRγ NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of RXRγ, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/nhr.
Catalog No. 50-200-0290
Only null left
Add to Cart
Request a Quote